Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084749.1 | NP_001078218.1 | 1 - 2 | 215 | 71 | |
NM_113690.3 | NP_566824.1 | 1 - 2 | 284 | 71 | ||
GenBank mRNA | AY046004.1 | - | 1 - 2 | 215 | 71 | |
AY079315.1 | - | 1 - 2 | 141 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |