Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338928.1 | NP_001319661.1 | 7 - 8 | 1039 | 64 | |
NM_001338929.1 | NP_001325749.1 | 7 - 8 | 1039 | 64 | ||
NM_001338930.1 | NP_001325748.1 | 6 - 7 | 1043 | 64 | ||
NM_001338931.1 | NP_001325750.1 | 4 - 5 | 493 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |