Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLC04 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113737.3 | NP_566835.1 | 1 - 2 | 1040 | 66 | |
GenBank mRNA | AY056286.1 | - | 1 - 2 | 922 | 66 | |
AY091433.1 | - | 1 - 2 | 641 | 66 | ||
KJ138809.1 | - | 1 - 2 | 641 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |