Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339023.1 | NP_001326115.1 | 8 - 9 | 930 | 74 | |
NM_113865.4 | NP_566850.3 | 8 - 9 | 812 | 74 | ||
GenBank mRNA | AY940684.1 | - | 8 - 9 | 772 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |