Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114433.3 | NP_190150.1 | 3 - 4 | 770 | 143 | |
GenBank mRNA | AF386961.1 | - | 3 - 4 | 571 | 143 | |
AK226663.1 | - | 574 | 143 | |||
BT000007.1 | - | 3 - 4 | 423 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |