Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035737.2 | NP_001030814.1 | 2 - 3 | 1091 | 74 | |
NM_114447.4 | NP_190164.1 | 3 - 4 | 1089 | 74 | ||
GenBank mRNA | AF030864.1 | - | 3 - 4 | 944 | 74 | |
AF360218.1 | - | 2 - 3 | 898 | 74 | ||
AY040062.1 | - | 2 - 3 | 881 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |