Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339237.1 | NP_001327061.1 | 2 - 3 | 1158 | 104 | |
NM_001339238.1 | NP_001327062.1 | 3 - 4 | 1080 | 104 | ||
NM_001339239.1 | NP_001327064.1 | 3 - 4 | 1080 | 104 | ||
NM_001339240.1 | NP_001327065.1 | 2 - 3 | 336 | 104 | ||
NM_001339241.1 | NP_001327063.1 | 2 - 3 | 315 | 104 | ||
NM_114489.5 | NP_190206.2 | 3 - 4 | 1080 | 104 | ||
GenBank mRNA | AY070078.1 | - | 3 - 4 | 237 | 104 | |
AY117353.1 | - | 3 - 4 | 220 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |