Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114564.3 | NP_190281.1 | 13 - 14 | 2353 | 71 | |
GenBank mRNA | AY090236.1 | - | 13 - 14 | 2074 | 71 | |
BT003012.1 | - | 12 - 13 | 1942 | 71 | ||
KJ138797.1 | - | 12 - 13 | 1942 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |