Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHT3;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114744.4 | NP_190454.1 | 2 - 3 | 836 | 89 | |
GenBank mRNA | BT033111.1 | - | 2 - 3 | 687 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |