Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114997.2 | NP_190706.1 | 1 - 2 | 543 | 77 | |
GenBank mRNA | BT029478.1 | - | 1 - 2 | 154 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |