Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117145.3 | NP_192815.1 | 3 - 4 | 749 | 122 | |
GenBank mRNA | AY045668.1 | - | 3 - 4 | 673 | 122 | |
BT004519.1 | - | 3 - 4 | 590 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |