Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117193.3 | NP_192861.1 | 3 - 4 | 1028 | 94 | |
GenBank mRNA | AY034901.1 | - | 3 - 4 | 650 | 94 | |
AY048273.1 | - | 3 - 4 | 637 | 94 | ||
AY057606.1 | - | 3 - 4 | 571 | 94 | ||
AY086294.1 | - | 3 - 4 | 787 | 94 | ||
BT002368.1 | - | 3 - 4 | 570 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |