Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCT1;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341357.1 | NP_001329436.1 | 2 - 3 | 223 | 137 | |
NM_118076.3 | NP_193691.2 | 4 - 5 | 596 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |