Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341906.1 | NP_001329053.1 | 2 - 3 | 1207 | 112 | |
NM_118962.5 | NP_567801.1 | 2 - 3 | 1207 | 112 | ||
GenBank mRNA | BT025339.1 | - | 2 - 3 | 339 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |