Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122048.4 | NP_568394.2 | 4 - 5 | 1374 | 84 | |
GenBank mRNA | AJ000331.1 | - | 4 - 5 | 1160 | 84 | |
KJ138675.1 | - | 4 - 5 | 1072 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |