Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122264.5 | NP_197748.1 | 4 - 5 | 1295 | 94 | |
GenBank mRNA | AK229078.1 | - | 4 - 5 | 1168 | 94 | |
BT026382.1 | - | 4 - 5 | 1109 | 94 | ||
U20388.1 | - | 4 - 5 | 1146 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |