Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122314.3 | NP_197797.1 | 1 - 2 | 403 | 60 | |
GenBank mRNA | BT020526.1 | - | 1 - 2 | 403 | 60 | |
BT021931.1 | - | 1 - 2 | 304 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |