Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161263.1 | NP_001154735.1 | 7 - 8 | 1036 | 113 | |
NM_001161264.1 | NP_001154736.1 | 7 - 8 | 1169 | 113 | ||
NM_001161265.1 | NP_001154737.1 | 7 - 8 | 1157 | 113 | ||
NM_001161266.1 | NP_001154738.1 | 7 - 8 | 1145 | 113 | ||
NM_001161267.1 | NP_001154739.1 | 7 - 8 | 1181 | 113 | ||
NM_001343885.1 | NP_001330963.1 | 7 - 8 | 1139 | 113 | ||
NM_001343886.1 | NP_001330964.1 | 6 - 7 | 1054 | 113 | ||
NM_001343887.1 | NP_001330968.1 | 6 - 7 | 1043 | 113 | ||
NM_001343888.1 | NP_001330967.1 | 6 - 7 | 1286 | 113 | ||
NM_001343889.1 | NP_001330966.1 | 5 - 6 | 1160 | 113 | ||
NM_001343890.1 | NP_001330965.1 | 7 - 8 | 1151 | 113 | ||
NM_122371.5 | NP_197851.5 | 7 - 8 | 1160 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |