Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332600.1 | NP_001320962.1 | 5 - 6 | 643 | 70 | |
NM_102191.6 | NP_564194.1 | 5 - 6 | 602 | 70 | ||
GenBank mRNA | AB493475.1 | - | 5 - 6 | 517 | 70 | |
AF195047.1 | - | 5 - 6 | 544 | 70 | ||
BT026410.1 | - | 5 - 6 | 517 | 70 | ||
DQ446281.1 | - | 5 - 6 | 517 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |