Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160892.1 | NP_001154364.1 | 4 - 5 | 665 | 97 | |
NM_102294.7 | NP_173858.5 | 5 - 6 | 815 | 97 | ||
GenBank mRNA | AM177312.1 | - | 5 - 6 | 776 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |