Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102595.2 | NP_564304.1 | 1 - 2 | 538 | 84 | |
GenBank mRNA | DQ446296.1 | - | 1 - 2 | 500 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |