Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036891.2 | NP_001031968.1 | 10 - 11 | 1331 | 71 | |
NM_001203493.1 | NP_001190422.1 | 10 - 11 | 1383 | 71 | ||
NM_122927.4 | NP_198386.1 | 10 - 11 | 1385 | 71 | ||
GenBank mRNA | AY085968.1 | - | 10 - 11 | 1328 | 71 | |
AY142630.1 | - | 10 - 11 | 1189 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |