Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123524.4 | NP_198975.1 | 3 - 4 | 730 | 92 | |
GenBank mRNA | AY035169.1 | - | 3 - 4 | 583 | 92 | |
AY057638.1 | - | 3 - 4 | 629 | 92 | ||
AY150449.1 | - | 3 - 4 | 529 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |