Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123726.4 | NP_199173.2 | 3 - 4 | 581 | 88 | |
GenBank mRNA | AK230338.1 | - | 1547 | 88 | ||
AY074343.1 | - | 3 - 4 | 545 | 88 | ||
AY091387.1 | - | 3 - 4 | 370 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |