Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT5G45430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203544.2 | NP_001190473.1 | 4 - 5 | 1191 | 153 | |
NM_123911.2 | NP_568646.1 | 4 - 5 | 953 | 153 | ||
GenBank mRNA | BT001210.1 | - | 1 - 2 | 84 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |