Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123965.2 | NP_199409.1 | 2 - 3 | 406 | 81 | |
GenBank mRNA | U43026.1 | - | 2 - 3 | 212 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |