Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124151.3 | NP_199588.1 | 11 - 12 | 1212 | 80 | |
GenBank mRNA | AF370324.1 | - | 11 - 12 | 1149 | 80 | |
AY063112.1 | - | 11 - 12 | 1025 | 80 | ||
U80605.1 | - | 11 - 12 | 1052 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |