Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124422.4 | NP_199851.2 | 3 - 4 | 615 | 83 | |
GenBank mRNA | AY842027.1 | - | 3 - 4 | 325 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |