Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124692.3 | NP_200125.1 | 6 - 7 | 1604 | 73 | |
GenBank mRNA | AF370139.1 | - | 6 - 7 | 1604 | 73 | |
BT000991.1 | - | 5 - 6 | 1211 | 73 | ||
Y16327.1 | - | 6 - 7 | 1515 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |