Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203600.1 | NP_001190529.1 | 12 - 13 | 3446 | 75 | |
NM_001203601.1 | NP_001190530.1 | 12 - 13 | 3446 | 75 | ||
NM_124725.5 | NP_200158.2 | 12 - 13 | 3461 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |