Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124796.4 | NP_200227.1 | 2 - 3 | 893 | 92 | |
GenBank mRNA | AY062837.1 | - | 2 - 3 | 798 | 92 | |
AY081565.1 | - | 2 - 3 | 720 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |