Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336557.1 | NP_001323748.1 | 5 - 6 | 2408 | 112 | |
NM_001336558.1 | NP_001323747.1 | 6 - 7 | 2258 | 112 | ||
NM_129103.5 | NP_565806.1 | 6 - 7 | 2118 | 112 | ||
GenBank mRNA | AY150402.1 | - | 4 - 5 | 1596 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |