Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336645.1 | NP_001325069.1 | 20 - 21 | 3557 | 88 | |
NM_001336646.1 | NP_001325068.1 | 19 - 20 | 3578 | 88 | ||
NM_001336647.1 | NP_001325070.1 | 19 - 20 | 3559 | 88 | ||
NM_129284.2 | NP_181265.1 | 20 - 21 | 3557 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |