Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHT4;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336686.1 | NP_001325125.1 | 6 - 7 | 1604 | 63 | |
NM_129362.4 | NP_181341.2 | 6 - 7 | 1612 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |