Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129385.5 | NP_181363.1 | 4 - 5 | 1226 | 69 | |
NM_201905.1 | NP_973634.1 | 3 - 4 | 1167 | 69 | ||
GenBank mRNA | AK226212.1 | - | 4 - 5 | 1026 | 69 | |
BT025321.1 | - | 4 - 5 | 996 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |