Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336920.1 | NP_001318403.1 | 4 - 5 | 1417 | 147 | |
NM_129714.4 | NP_181682.2 | 4 - 5 | 1537 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |