Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148142.3 | NP_680447.1 | 11 - 12 | 1471 | 59 | |
GenBank mRNA | AK118919.1 | - | 11 - 12 | 1402 | 59 | |
AY070458.1 | - | 11 - 12 | 1309 | 59 | ||
AY091321.1 | - | 11 - 12 | 991 | 59 | ||
AY943855.1 | - | 11 - 12 | 991 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |