Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335662.1 | NP_001324133.1 | 2 - 3 | 338 | 180 | |
NM_127570.5 | NP_565464.2 | 1 - 2 | 370 | 180 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |