Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338231.1 | NP_001327031.1 | 2 - 3 | 299 | 130 | |
NM_112510.3 | NP_566545.1 | 1 - 2 | 245 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |