Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQ14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125450.2 | NP_001118922.1 | 2 | 1216 | 148 | |
NM_116523.3 | NP_567247.2 | 3 | 850 | 148 | ||
NM_178960.2 | NP_849291.1 | 3 | 853 | 148 | ||
NM_178961.1 | NP_849292.1 | 1 | 1375 | 148 |
Gene Symbol: UBQ10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK316842.1 | - | 2 | 1079 | 148 | |
AY087759.1 | - | 2 | 1079 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |