Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340172.1 | NP_001327941.1 | 10 - 11 | 1208 | 151 | |
NM_001340174.1 | NP_001327939.1 | 10 - 11 | 1166 | 151 | ||
NM_001340175.1 | NP_001327940.1 | 9 - 10 | 1238 | 151 | ||
NM_001340176.1 | NP_001327937.1 | 9 - 10 | 1184 | 151 | ||
NM_116140.4 | NP_191834.3 | 10 - 11 | 1136 | 151 | ||
GenBank mRNA | AK175256.1 | - | 10 - 11 | 1085 | 151 | |
AK176786.1 | - | 10 - 11 | 1084 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |