Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083981.2 | NP_001077450.1 | 1 - 2 | 221 | 90 | |
GenBank mRNA | AK176405.1 | - | 1 - 2 | 164 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |