Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076095.1 | NP_001069563.1 | 1 - 2 | 371 | 95 | |
GenBank mRNA | BC118177.1 | - | 1 - 2 | 371 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |