Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034394.2 | NP_001029566.1 | 3 - 4 | 704 | 114 | |
GenBank mRNA | BC102922.1 | - | 3 - 4 | 705 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |