Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174032.2 | NP_776457.1 | 2 - 3 | 208 | 65 | |
GenBank mRNA | AB099005.1 | - | 2 - 3 | 203 | 65 | |
BC102312.1 | - | 2 - 3 | 254 | 65 | ||
BC151425.1 | - | 2 - 3 | 208 | 65 | ||
BT021022.1 | - | 2 - 3 | 184 | 65 | ||
X91755.1 | - | 208 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |