Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174102.2 | NP_776527.1 | 4 - 5 | 524 | 57 | |
GenBank mRNA | L04188.1 | - | 4 - 5 | 524 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |