Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCKDHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174506.1 | NP_776931.1 | 8 - 9 | 1292 | 60 | |
GenBank mRNA | BC109669.1 | - | 8 - 9 | 1186 | 60 | |
J03759.1 | - | 8 - 9 | 1292 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |