Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174515.2 | NP_776940.1 | 5 - 6 | 468 | 55 | |
GenBank mRNA | BC149340.1 | - | 5 - 6 | 496 | 55 | |
X93604.1 | - | 468 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |