Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GKN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001148.1 | NP_001001148.1 | 3 - 4 | 250 | 78 | |
GenBank mRNA | AY456959.1 | - | 3 - 4 | 250 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |