Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015587.1 | NP_001015587.1 | 4 - 5 | 505 | 89 | |
GenBank mRNA | BC126566.1 | - | 4 - 5 | 645 | 89 | |
BT020902.1 | - | 4 - 5 | 505 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |